missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Propyl Sulfide 95.0+%, TCI America™

Catalog No. p-7134794
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08265ML 5 mL
P082625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08265ML Supplier TCI America Supplier No. P08265ML

Chemical Identifiers

CAS 5008-73-1
Molecular Formula C6H14S
Molecular Weight (g/mol) 118.238
MDL Number MFCD00027125
InChI Key BDFDQOJJDDORSR-UHFFFAOYSA-N
PubChem CID 78708
IUPAC Name 1-propan-2-ylsulfanylpropane
SMILES CCCSC(C)C

Specifications

Color Yellow
Boiling Point 132°C
Quantity 5 mL
UN Number 1993
Formula Weight 118.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Propyl Sulfide