missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Palmitate 97.0+%, TCI America™

Supplier:  TCI America P0005500ML

 View more versions of this product

Catalog No. P0005500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Isopropyl Palmitate
13°C
160°C
MFCD00008993
isopropyl palmitate, isopropyl hexadecanoate, isopalm, deltyl, isopal, propal, deltyl prime, tegester isopalm, ja-fa ippkessco, sinnoester pit
CCCCCCCCCCCCCCCC(=O)OC(C)C
298.51
CHEBI:84262
≥97.0% (GC)
142-91-6
Yellow
C19H38O2
500 mL
XUGNVMKQXJXZCD-UHFFFAOYSA-N
propan-2-yl hexadecanoate
8907
298.51
Liquid

Chemical Identifiers

142-91-6
298.51
XUGNVMKQXJXZCD-UHFFFAOYSA-N
8907
propan-2-yl hexadecanoate
C19H38O2
MFCD00008993
isopropyl palmitate, isopropyl hexadecanoate, isopalm, deltyl, isopal, propal, deltyl prime, tegester isopalm, ja-fa ippkessco, sinnoester pit
CHEBI:84262
CCCCCCCCCCCCCCCC(=O)OC(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0798

RTECSNumber : RT4900000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions