accessibility menu, dialog, popup

UserName

H3COOCH3CH3

CAS RN 142-91-6

IUPAC Name: propan-2-yl hexadecanoate
Synonyms: isopropyl palmitate estol 103 plymouth ipp JA-FA ipp isopalm nikkol ipp ja-fa ippkessco emerest 2316 wickenol 111 isopal
Molecular Weight (g/mol): 298.51
Molecular Formula: C19H38O2
InChi Key: XUGNVMKQXJXZCD-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC(=O)OC(C)C
Molecular Weight (g/mol) 
  • (3)
  • (11)
Percent Purity 
  • (3)
  • (2)
  • (3)
  • (6)
Grade 
  • (1)
  • (9)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (9)
  • (3)
  • (1)
  • (6)
  • (2)
  • (3)
  • (3)
  • (11)
  • (3)
  • (2)
  • (3)
  • (6)
  • (1)
  • (9)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (7)
  • (2)
  • (3)