missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Palmitate 97.0+%, TCI America™

Catalog No. p-7134369
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
P000525ML 25 mL
P0005500ML 500 mL
2 options

Catalog No. P000525ML

Supplier: TCI America P000525ML

Only null left

Chemical Identifiers

CAS 142-91-6
Molecular Formula C19H38O2
Molecular Weight (g/mol) 298.51
MDL Number MFCD00008993
InChI Key XUGNVMKQXJXZCD-UHFFFAOYSA-N
Synonym isopropyl palmitate, isopropyl hexadecanoate, isopalm, deltyl, isopal, propal, deltyl prime, tegester isopalm, ja-fa ippkessco, sinnoester pit
PubChem CID 8907
ChEBI CHEBI:84262
IUPAC Name propan-2-yl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OC(C)C

Specifications

Melting Point 13°C
Color Yellow
Boiling Point 160°C
Quantity 25 mL
Formula Weight 298.51
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Palmitate
EINECSNumber (2)-0798
RTECSNumber RT4900000
TSCA Yes