missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl chloroacetate, 99%

CAS: 105-48-6 | C5H9ClO2 | 136.58 g/mol

$53.32 - $53.32

Chemical Identifiers

CAS 105-48-6
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.58
MDL Number MFCD00040410
InChI Key VODRWDBLLGYRJT-UHFFFAOYSA-N
Synonym isopropyl chloroacetate, chloroacetic acid isopropyl ester, acetic acid, chloro-, 1-methylethyl ester, iso-propyl chloroacetate, clch2c o och ch3 2, ccris 7748, acetic acid, chloro-, isopropyl ester, acetic acid, 2-chloro-, 1-methylethyl ester, chloroacetic acid, 1-methyl ester, monochloroacetic acid isopropyl ester
PubChem CID 7759
IUPAC Name propan-2-yl 2-chloroacetate
SMILES CC(C)OC(=O)CCl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA19070AC
View Documents
Thermo Scientific Chemicals
A19070AC
25 mL
Each for $53.32
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

105-48-6
136.58
VODRWDBLLGYRJT-UHFFFAOYSA-N
7759
CC(C)OC(=O)CCl
C5H9ClO2
MFCD00040410
isopropyl chloroacetate, chloroacetic acid isopropyl ester, acetic acid, chloro-, 1-methylethyl ester, iso-propyl chloroacetate, clch2c o och ch3 2, ccris 7748, acetic acid, chloro-, isopropyl ester, acetic acid, 2-chloro-, 1-methylethyl ester, chloroacetic acid, 1-methyl ester, monochloroacetic acid isopropyl ester
propan-2-yl 2-chloroacetate

Specifications

105-48-6
149°C to 150°C
1.4205
MFCD00040410
UN2947
VODRWDBLLGYRJT-UHFFFAOYSA-N
propan-2-yl 2-chloroacetate
7759
99%
1.08
C5H9ClO2
ClCH2CO2CH(CH3)2
25 mL
isopropyl chloroacetate, chloroacetic acid isopropyl ester, acetic acid, chloro-, 1-methylethyl ester, iso-propyl chloroacetate, clch2c o och ch3 2, ccris 7748, acetic acid, chloro-, isopropyl ester, acetic acid, 2-chloro-, 1-methylethyl ester, chloroacetic acid, 1-methyl ester, monochloroacetic acid isopropyl ester
CC(C)OC(=O)CCl
136.58
136.58
Isopropyl chloroacetate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H301-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ISOPROPYL CHLOROACETATE

EINECSNumber : 203-301-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only