accessibility menu, dialog, popup

UserName

H3CCH3OOCl

CAS RN 105-48-6

IUPAC Name: propan-2-yl 2-chloroacetate
Synonyms: isopropyl chloroacetate
Molecular Weight (g/mol): 136.58
Molecular Formula: C5H9ClO2
InChi Key: VODRWDBLLGYRJT-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)CCl
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)