missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, Technical, Spectrum™ Chemical

$254.29 - $8140.43

Chemical Identifiers

CAS 67-63-0
Molecular Weight (g/mol) 60.1
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18611132
View Documents
Spectrum Chemical Mfg Cor
I1051500MLGL
500 mL Amber Glass Bottle
Each for $254.29
Only null left
 
18611139
View Documents
Spectrum Chemical Mfg Cor
I10514LTGL
4 L Amber Glass Bottle
Each for $466.86
Only null left
 
18611140
View Documents
Spectrum Chemical Mfg Cor
I10514LTPL
4 L Poly Bottle
Each for $509.39
Only null left
 
18611133
View Documents
Spectrum Chemical Mfg Cor
I105120LTMT
20 L Steel Can
Each for $1,728.93
Only null left
 
18611131
View Documents
Spectrum Chemical Mfg Cor
I1051200LTBL
200 L Drum
Each for $8,140.43
Only null left
 
Description

Description

Specifications

Chemical Identifiers

67-63-0
60.1

Specifications

67-63-0
78.3°C
Amber Glass Bottle
Technical
1
500 mL
60.1
Isopropyl Alcohol
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.