missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Alcohol Solution, LC-MS, For flushing LC-MS systems, 50/50 Isopropyl Alcohol / Water, Honeywell Burdick & Jackson
Supplier: Honeywell-Burdick and Jackson LC7034
Specifications
| Isopropyl Alcohol Solution | |
| 67-63-0,7732-18-5 | |
| 55.6000%,44.4000% | |
| UN1219 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CC(C)O | |
| 60.096 | |
| CHEBI:17824 | |
| Glass Bottle | |
| 22.78°C |
| LC-MS, For flushing LC-MS systems, 50/50 Isopropyl Alcohol / Water | |
| 82.3°C | |
| C3H8O | |
| 0.9189 g/cm3 | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| propan-2-ol | |
| 3776 | |
| LC-MS | |
| 4 x 4 L |
Chemical Identifiers
| 67-63-0 | |
| 60.096 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CHEBI:17824 | |
| CC(C)O |
| C3H8O | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| 3776 | |
| propan-2-ol |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Isopropanol solution