missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Alcohol, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.9%, Honeywell Burdick & Jackson
$1180.53 - $10555.23
Chemical Identifiers
| CAS | 67-63-0 |
|---|---|
| Molecular Formula | C3H8O |
| Molecular Weight (g/mol) | 60.096 |
| MDL Number | MFCD00064589 |
| InChI Key | KFZMGEQAYNKOFK-UHFFFAOYSA-N |
| Synonym | isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
| PubChem CID | 3776 |
| ChEBI | CHEBI:17824 |
| IUPAC Name | propan-2-ol |
| SMILES | CC(C)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
6000157
|
Honeywell-Burdick and Jackson
PP32319 |
19 L | Metal Can |
Each for $1,180.53
|
|
||||
|
6000158
|
Honeywell Chemicals
PP323204 |
204 L | Metal Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 67-63-0 | |
| 60.096 | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| 3776 | |
| propan-2-ol |
| C3H8O | |
| MFCD00064589 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CHEBI:17824 | |
| CC(C)O |
Specifications
| 67-63-0 | |
| 0.785 g/cm3 | |
| 19 L | |
| Metal Can | |
| H2SO4 | |
| UN1219 | |
| 635639 | |
| Miscible in Water at 20°C | |
| CC(C)O | |
| 60.096 | |
| CHEBI:17824 | |
| >99.9% | |
| Isopropyl Alcohol, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.9% |
| 100.0000% | |
| 82.3°C | |
| 12°C | |
| C3H8O | |
| MFCD00064589 | |
| Passes test | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| propan-2-ol | |
| 3776 | |
| 60.11 | |
| ACS Reagent | |
| by GC Analysis |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Isopropanol
EINECSNumber : 200-661-7