missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.9%, Honeywell Burdick & Jackson

Catalog No. 6000158
Change view
Click to view available options
Quantity:
19 L
204 L
Packaging:
Metal Can
Metal Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000158 204 L Metal Drum
6000157 19 L Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000158 Supplier Honeywell Chemicals Supplier No. PP323204

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00064589
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

CAS Max % 100.0000%
Density 0.785 g/cm3
Quantity 204 L
Flash Point 12°C
Packaging Metal Drum
Linear Formula H2SO4
UN Number UN1219
Water Passes test
Beilstein 635639
Boiling Point 82.3°C
Solubility Information Miscible in Water at 20°C
Formula Weight 60.11
Percent Purity >99.9%
Grade ACS
Assay by GC Analysis
Chemical Name or Material Isopropyl Alcohol, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Isopropanol
EINECSNumber 200-661-7