missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, Histological Grade, Spectrum™ Chemical

$907.00 - $11237.86

Chemical Identifiers

CAS 67-63-0
Molecular Weight (g/mol) 60.1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18611121
View Documents
Spectrum Chemical Mfg Cor
HS0044LTGL
4 L Amber Glass Bottle
Each for $907.00
Only null left
 
18611122
View Documents
Spectrum Chemical Mfg Cor
HS00420LT
20 L Amber Glass Bottle
Each for $2,221.46
Only null left
 
18611123
View Documents
Spectrum Chemical Mfg Cor
HS004200LTBL
200 L Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

67-63-0
60.1

Specifications

Liquid
1
Colorless
60.1
Isopropyl Alcohol
67-63-0
Amber Glass Bottle
4 L
Histological
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.