missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, Histological Grade, Spectrum™ Chemical

Catalog No. 18611123
Change view
Click to view available options
Quantity:
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611123 200 L Amber Glass Bottle
18611121 4 L Amber Glass Bottle
18611122 20 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18611123 Supplier Spectrum Chemical Mfg Cor Supplier No. HS004200LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-63-0
Molecular Weight (g/mol) 60.1

Specifications

Physical Form Liquid
CAS Max % 1
Packaging Amber Glass Bottle
Color Colorless
Quantity 200 L
Grade Histological
Chemical Name or Material Isopropyl Alcohol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.