missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, FCC, 99%, Spectrum™ Chemical

I1056, 67-63-0, C3H8O

$394.57 - $4,060.82

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
IUPAC Name propan-2-ol
SMILES CC(C)O
View More Specs

Products 10
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18603515
View Documents
Spectrum Chemical Mfg Cor
I1056500MLGL
500 mL Amber Glass Bottle N/A
Only null left
 
18603517
View Documents
Spectrum Chemical Mfg Cor
I10561LT
1 L Amber Glass Bottle N/A
Only null left
 
18603516
View Documents
Spectrum Chemical Mfg Cor
I1056500MLGLCS6
6 x 500 mL Amber Glass Bottle N/A
Only null left
 
18603518
View Documents
Spectrum Chemical Mfg Cor
I10564LTGL
4 L Amber Glass Bottle N/A
Only null left
 
18603519
View Documents
Spectrum Chemical Mfg Cor
I10564LTPL
4 L Poly Bottle N/A
Only null left
 
18603520
View Documents
Spectrum Chemical Mfg Cor
I10561LTCS6
6 x 1 L Amber Glass Bottle N/A
Only null left
 
18603521
View Documents
Spectrum Chemical Mfg Cor
I10564LTGLCS4
4 x 4 L Amber Glass Bottle N/A
Only null left
 
18603522
View Documents
Spectrum Chemical Mfg Cor
I10564LTPLCS4
4 x 4 L Poly Bottle N/A
Only null left
 
18603523
View Documents
Spectrum Chemical Mfg Cor
I105620LT
20 L Steel Can N/A
Only null left
 
18603524
View Documents
Spectrum Chemical Mfg Cor
I1056200LTBL
200 L Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Isopropyl Alcohol, 99 Percent, FCC is used to sanitize food processing equipment. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

67-63-0
60.10
propan-2-ol
C3H8O
KFZMGEQAYNKOFK-UHFFFAOYSA-N
CC(C)O

Specifications

67-63-0
100%
500 mL
0.784
UN1219
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
99.7%
Isopropyl Alcohol
10 mg/kg
78.3°C
Amber Glass Bottle
C3H8O
0.2%
CC(C)O
60.10
FCC
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'