missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, FCC, 99%, Spectrum™ Chemical

I1056, 67-63-0, C3H8O

Supplier:  Spectrum Chemical Mfg Cor I10564LTPLCS4

 View more versions of this product

Catalog No. 18603522


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Isopropyl Alcohol, 99 Percent, FCC is used to sanitize food processing equipment. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

Isopropyl Alcohol
10 mg/kg
78.3°C
Poly Bottle
C3H8O
0.2%
CC(C)O
60.10
FCC
67-63-0
100%
4 x 4 L
0.784
UN1219
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
99.7%

Chemical Identifiers

67-63-0
60.10
propan-2-ol
C3H8O
KFZMGEQAYNKOFK-UHFFFAOYSA-N
CC(C)O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions