missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, B&J Brand™, for HPLC, GC, pesticide residue analysis, and spectrophotometry, >99.9%, Honeywell Burdick & Jackson

$4745.05 - $4745.05

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
6000147
View Documents
Honeywell Chemicals
CS32356
56 L Returnable Steel Drum
Each for $4,745.05
Only null left
 
Description

Description

Specifications

Chemical Identifiers

67-63-0
60.096
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CHEBI:17824
CC(C)O
C3H8O
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
propan-2-ol

Specifications

67-63-0
100.0000%
82.3°C
12°C
C3H8O
0.06% max.
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
3776
60.11
HPLC
0.16 AU max. at 225 nm, 0.02 AU max. at 250 nm, 0.005 AU max. at 300 nm, 0.005 AU max. at 400 nm
0.785 g/cm3
56 L
Returnable Steel Drum
UN1219
635639
Miscible in Water at 20°C
CC(C)O
60.096
CHEBI:17824
>99.9%
Isopropyl Alcohol, B&J Brand™, for HPLC, GC, pesticide residue analysis, and spectrophotometry, >99.9%
Videos
Safety and Handling

Safety and Handling

ShelfLife : 547 days from date of manufacture

DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Isopropanol

SDS
missing translation for 'documents'

missing translation for 'documents'