missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Alcohol, ACS Grade, MilliporeSigma™
Supplier: Technidata PX18352
Specifications
| Isopropyl Alcohol | |
| 67-63-0 | |
| 0.78g/cm3 at 20°C | |
| 1 L | |
| 399°C (750.2°F) | |
| Glass Bottle | |
| C3H8O | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| propan-2-ol | |
| 3776 | |
| 60.1 | |
| ACS Reagent |
| -88.9°C | |
| Colorless | |
| 82.4°C | |
| 11.85°C (53.3°F) | |
| Charactristic | |
| Liquid | |
| UN 1219 | |
| Soluble in water | |
| CC(C)O | |
| 60.096 | |
| CHEBI:17824 | |
| ≥99.5% |
Chemical Identifiers
| 67-63-0 | |
| 60.096 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CHEBI:17824 | |
| CC(C)O |
| C3H8O | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| 3776 | |
| propan-2-ol |
Safety and Handling
DOTInformation : DOT Class: 3, Packaging Group: II
Recommended Storage : Room Temperature