missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, ACS Grade, MilliporeSigma™

Catalog No. p-3993668
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Poly-coated Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MPX18352 1 L Glass Bottle
MPX1835P4 4 L Poly-coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MPX18352 Supplier Technidata Supplier No. PX18352

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -88.9°C
Color Colorless
Density 0.78g/cm3 at 20°C
Boiling Point 82.4°C
Quantity 1 L
Flash Point 11.85°C (53.3°F)
Ignition Point 399°C (750.2°F)
Odor Charactristic
Packaging Glass Bottle
Physical Form Liquid
UN Number UN 1219
Solubility Information Soluble in water
Formula Weight 60.1
Percent Purity ≥99.5%
Grade ACS Reagent
Chemical Name or Material Isopropyl Alcohol
Show More Show Less
DOTInformation DOT Class: 3, Packaging Group: II
Recommended Storage Room Temperature