missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Alcohol, ACS Grade, ≥ 99.5%, LabChem™
Supplier: LabChem LC1575044
Specifications
| Isopropyl Alcohol | |
| 67-63-0 | |
| 100 | |
| 785kg/m3 | |
| 4 x 4 L | |
| Passes Test | |
| Liquid | |
| 785kg/m3 | |
| C3H8O | |
| UN1219 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| propan-2-ol | |
| 3776 | |
| 60.09 | |
| ACS |
| -88°C | |
| Colorless | |
| Carbon dioxide; Carbon monoxide | |
| 82°C | |
| 12°C | |
| Poly (HDPE) bottle | |
| ≤0.001% | |
| C3H8O | |
| 2.5316mm2/s | |
| ≤0.2% | |
| Soluble in water | |
| CC(C)O | |
| 60.096 | |
| CHEBI:17824 | |
| ≥99.5% |
Chemical Identifiers
| 67-63-0 | |
| 60.096 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CHEBI:17824 | |
| CC(C)O |
| C3H8O | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| 3776 | |
| propan-2-ol |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Danger
EINECSNumber : 200-661-7
Recommended Storage : Room Temperature