missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, ACS Grade, ≥ 99.5%, LabChem™

Catalog No. p-5379506
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 x 4 L
Packaging:
Cubitainer
Poly (HDPE) bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC157501 500 mL Poly (HDPE) bottle
LC157502 1 L Poly (HDPE) bottle
LC157504 4 L Poly (HDPE) bottle
LC1575044 4 x 4 L Poly (HDPE) bottle
LC157505 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. LC157501 Supplier LabChem Supplier No. LC157501
Only null left

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -88°C
Color Colorless
CAS Max % 100
Decomposition Information Carbon dioxide; Carbon monoxide
Density 785kg/m3
Quantity 500 mL
Flash Point 12°C
Identification Passes Test
Packaging Poly (HDPE) bottle
Physical Form Liquid
Residue after Evaporation ≤0.001%
Specific Gravity 785kg/m3
Linear Formula C3H8O
Viscosity 2.5316mm2/s
UN Number UN1219
Water ≤0.2%
Solubility Information Soluble in water
Boiling Point 82°C
Formula Weight 60.09
Percent Purity ≥99.5%
Grade ACS
Chemical Name or Material Isopropyl Alcohol
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Precautionary Statement GHS P Statement
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Avoid breathing mist, spray, vapors.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Call a poison center/doctor if you feel unwell.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use dry chemical powder, alcohol-resistant foam, carbon dioxide to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 200-661-7
Recommended Storage Room Temperature