missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Alcohol, 90% (v/v), Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 9.0, 89.0 |
| CAS Max % | 11.0, 91.0 |
| Boiling Point | 82°C |
| Quantity | 1 L |
| Packaging | Natural Poly Bottle |
| Physical Form | Liquid |
| Percent Purity | 90% |
| Grade | Laboratory |
| Concentration or Composition (by Analyte or Components) | 90% (v/v) |
| Show More |