missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, 50% v/v, Reagents

Catalog No. 01334901
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334901 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334901 Supplier Labchem Inc Supplier No. CI2963001A
Only null left

Isopropyl Alcohol, 50% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 67-63-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Isopropyl Alcohol, 2-Propanol, Propan-2-ol
IUPAC Name water
SMILES O

Specifications

Color Colorless
Quantity 1 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 0.9257
Boiling Point 81°C
Percent Purity 50%
Grade Reagent
Chemical Name or Material Isopropyl Alcohol, 50% v/v