missing translation for 'onlineSavingsMsg'
Learn More

Isopropenylmagnesium bromide, 0.5M solution in THF, AcroSeal™

Catalog No. AC427758000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427758000 800 mL AcroSeal™ Glass Bottle
AC427751000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC427758000 Supplier Thermo Scientific Chemicals Supplier No. 427758000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13291-18-4 | C3H5BrMg | 145.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13291-18-4 , 109-99-9
Molecular Formula C3H5BrMg
Molecular Weight (g/mol) 145.28
MDL Number MFCD00192240
InChI Key LFTYALBVGVNGLI-UHFFFAOYSA-M
Synonym isopropenylmagnesium bromide, isopropenylmagnesium bromide solution, isopropenyl magnesium bromide, bromo prop-1-en-2-yl magnesium, magnesium, bromo 1-methylethenyl, bromo isopropenyl magnesium, 1-methylethenylmagnesium bromide, 2-propen-2-ylmagnesium bromide, prop-1-en-2-ylmagnesium bromide, lftyalbvgvngli-uhfffaoysa-m, grignard reagent
PubChem CID 4187787
IUPAC Name magnesium;prop-1-ene;bromide
SMILES CC(=C)[Mg]Br

Specifications

Color Orange to White
Density 0.9270g/mL
Boiling Point 67.0°C
Flash Point 2°C
Packaging AcroSeal™ Glass Bottle
Linear Formula CH2=C(CH3)MgBr
Quantity 800 mL
Specific Gravity 0.927
Formula Weight 145.28
Physical Form Crystalline Powder
Chemical Name or Material Isopropenylmagnesium bromide, 0.5M Solution in THF
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
May form explosive
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture

RUO – Research Use Only