CAS RN 13291-18-4
IUPAC Name:
bromo(prop-1-en-2-yl)magnesium
Synonyms:
bromo(prop-1-en-2-yl)magnesium
Molecular Weight (g/mol):
145.28
Molecular Formula:
C3H5BrMg
InChi Key:
LFTYALBVGVNGLI-UHFFFAOYSA-M
SMILES:
CC(=C)[Mg]Br
1
–
1
of
1
results
1
Promotions Available
Isopropenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
Isopropenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
CAS: 13291-18-4 Molecular Formula: C3H5BrMg Molecular Weight (g/mol): 145.28 MDL Number: MFCD00192240 InChI Key: LFTYALBVGVNGLI-UHFFFAOYSA-M Synonym: isopropenylmagnesium bromide,isopropenylmagnesium bromide solution,isopropenyl magnesium bromide,bromo prop-1-en-2-yl magnesium,magnesium, bromo 1-methylethenyl,bromo isopropenyl magnesium,1-methylethenylmagnesium bromide,2-propen-2-ylmagnesium bromide,prop-1-en-2-ylmagnesium bromide,lftyalbvgvngli-uhfffaoysa-m,grignard reagent PubChem CID: 4187787 IUPAC Name: magnesium;prop-1-ene;bromide SMILES: CC(=C)[Mg]Br