missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropanol/Water, 91:9, Reagents
Specifications
Specifications
| Color | Colorless |
| Quantity | 200 L |
| Packaging | Poly Drum |
| Physical Form | Liquid |
| Specific Gravity | 0.8159 |
| Boiling Point | 82°C |
| Percent Purity | 91% |
| Grade | Reagent |
| Chemical Name or Material | Isopropanol/Water, 91:9 |