Learn More
Isopropanol, 99.5%, for molecular biology, DNAse, RNAse and Protease free
CAS: 67-63-0 | C3H8O | 60.096 g/mol
Supplier: Thermo Scientific Chemicals 327270010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Isopropanol | |
| 67-63-0 | |
| 1 L | |
| Liquid | |
| 99.5% | |
| (CH3)2CHOH | |
| Vapor Pressure: 43mmHg at 20°C | |
| 01, 360 | |
| 15, 5254 | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| propan-2-ol | |
| 3776 | |
| 60.1 | |
| Molecular Biology |
| DNAse, RNAse and Protease Free | |
| 2.1 (Air = 1.0) | |
| Glass Bottle | |
| 0.0005% max. | |
| C3H8O | |
| MFCD00011674 | |
| 0.05% max. (Coulometric) | |
| 13,108 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CC(C)O | |
| 60.096 | |
| CHEBI:17824 | |
| ≥99.5% |
Chemical Identifiers
| 67-63-0 | |
| 60.096 | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| 3776 | |
| propan-2-ol |
| C3H8O | |
| MFCD00011674 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CHEBI:17824 | |
| CC(C)O |
Safety and Handling
GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Emergency Overview
Flammable liquid and vapor. Irritating to eyes and skin. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and
Signal Word: WARNING!
RUO – Research Use Only