missing translation for 'onlineSavingsMsg'
Learn More

Isopropanol, 99.5%, for molecular biology, DNAse, RNAse and Protease free

Catalog No. p-3737535
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC327272500 250 mL Glass Bottle
AC327270010 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC327272500 Supplier Thermo Scientific Chemicals Supplier No. 327272500
Only null left

CAS: 67-63-0 | C3H8O | 60.096 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Density 2.1 (Air = 1.0)
Quantity 250 mL
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range 99.5%
Residue after Evaporation 0.0005% max.
Linear Formula (CH3)2CHOH
Vapor Pressure Vapor Pressure: 43mmHg at 20°C
Beilstein 01, 360
Water 0.05% max. (Coulometric)
Fieser 13,108
Merck Index 15, 5254
Formula Weight 60.1
Percent Purity ≥99.5%
Grade Molecular Biology
Chemical Name or Material Isopropanol, DNAse, RNAse and Protease Free
Show More Show Less
Hazard Category Signal Word: WARNING!
Hazard Statement GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Irritating to eyes and skin. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and

RUO – Research Use Only