missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, 70% (v/v), Ricca Chemical

Catalog No. 42105
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
55 gal.
Packaging:
Cubitainer
Hard Pack
Natural Poly Bottle
Poly Drum
Safety-coated Glass Bottle
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
42105 20 L Cubitainer
421016 500 mL Natural Poly Bottle
421032 1 L Natural Poly Bottle
42101 4 L Natural Poly Bottle
R42100004C1 4 L Safety-coated Glass Bottle
421025 10 L Cubitainer
42105HP 20 L Hard Pack
421055 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 42105 Supplier Ricca Chemical Company Supplier No. 42105

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-63-0 , 7732-18-5
Molecular Formula C3H8O
Molecular Weight (g/mol) Mixture
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

CAS Min % 29.0, 69.0
CAS Max % 31.0, 71.0
Appearance Colorless Liquid
Quantity 20 L
Packaging Cubitainer
Specific Gravity 0.874 to 0.882
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
Boiling Point 82°C
Percent Purity 68 to 72% v/v
Grade Laboratory
Chemical Name or Material Isopropanol
Show More Show Less
ShelfLife 24 months