missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, 10% (v/v), Ricca Chemical

Catalog No. p-3251838
Change view
Click to view available options
Quantity:
4 L
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
42071 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42071 Supplier Ricca Chemical Company Supplier No. 42071
Only null left

Chemical Identifiers

CAS 7732-18-5 , 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) Mixture
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Color Colorless
CAS Min % 89.0, 9.0
CAS Max % 91.0, 11.0
Quantity 4 L
Packaging Natural Poly Bottle
Physical Form Liquid
Boiling Point 100°C
Percent Purity 10%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 10% (v/v)
Chemical Name or Material Isopropanol
Show More Show Less