missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Alcohol, 10% (v/v), Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 89.0, 9.0 |
| CAS Max % | 91.0, 11.0 |
| Quantity | 4 L |
| Packaging | Natural Poly Bottle |
| Physical Form | Liquid |
| Boiling Point | 100°C |
| Percent Purity | 10% |
| Grade | Laboratory |
| Concentration or Composition (by Analyte or Components) | 10% (v/v) |
| Show More |