missing translation for 'onlineSavingsMsg'
Learn More

Isopropamide Iodide 95.0+%, TCI America™

Catalog No. i10481g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I10481G 1 g
I10485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I10481G Supplier TCI America Supplier No. I10481G
Only null left

Chemical Identifiers

CAS 71-81-8
Molecular Formula C23H33IN2O
Molecular Weight (g/mol) 480.434
MDL Number MFCD00063513
InChI Key BFSMWENDZZIWPW-UHFFFAOYSA-N
Synonym 4-Amino-N,N-diisopropyl-N-methyl-4-oxo-3,3-diphenyl-1-butanaminium Iodide, (3-Carbamoyl-3,3-diphenylpropyl)diisopropylmethylammonium Iodide
PubChem CID 6284
IUPAC Name (4-amino-4-oxo-3,3-diphenylbutyl)-methyl-di(propan-2-yl)azanium;iodide
SMILES CC(C)[N+](C)(CCC(C1=CC=CC=C1)(C2=CC=CC=C2)C(=O)N)C(C)C.[I-]

Specifications

Melting Point 201°C
Color White
Quantity 1 g
Formula Weight 480.43
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isopropamide Iodide
EINECSNumber (9)-0178
TSCA No