accessibility menu, dialog, popup

UserName

CAS RN 71-81-8

IUPAC Name: (3-carbamoyl-3,3-diphenylpropyl)(methyl)bis(propan-2-yl)azanium iodide
Synonyms: isopropamide iodide
Molecular Weight (g/mol): 480.43
Molecular Formula: C23H33IN2O
InChi Key: BFSMWENDZZIWPW-UHFFFAOYSA-N
SMILES: [I-].CC(C)[N+](C)(CCC(C(N)=O)(C1=CC=CC=C1)C1=CC=CC=C1)C(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results