missing translation for 'onlineSavingsMsg'
Learn More

Isoprenaline hydrochloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 437210250

 View more versions of this product

Catalog No. AC437210250

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Isoprenaline hydrochloride
165.0°C to 175.0°C
Authentic
Glass bottle
MFCD00012603,MFCD00064548
isoprenaline hydrochloride, isoproterenol hydrochloride, isuprel, euspiran, isoprenaline hcl, isoproterenol hcl, aerolone, aerotrol, izadrin, vapo-iso
IROWCYIEJAOFOW-UHFFFAOYNA-N
4-[1-hydroxy-2-(propan-2-ylamino)ethyl]benzene-1,2-diol;hydrochloride
5807
98%
51-30-9
1% max.
97.5% min. (HPLC)
C11H18ClNO3
25 g
Solubility in water: soluble in cold water.
[H+].[Cl-].CC(C)NCC(O)C1=CC=C(O)C(O)=C1
247.72
247.72

Chemical Identifiers

51-30-9
247.72
IROWCYIEJAOFOW-UHFFFAOYNA-N
5807
[H+].[Cl-].CC(C)NCC(O)C1=CC=C(O)C(O)=C1
C11H18ClNO3
MFCD00012603,MFCD00064548
isoprenaline hydrochloride, isoproterenol hydrochloride, isuprel, euspiran, isoprenaline hcl, isoproterenol hcl, aerolone, aerotrol, izadrin, vapo-iso
4-[1-hydroxy-2-(propan-2-ylamino)ethyl]benzene-1,2-diol;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED:

GHS Signal Word: Warning

EINECSNumber : 200-089-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only