missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isophthalaldehyde, 98%
Description
Isophthalaldehyde is used in the synthesis of binuclear ruthenium complex. It participates in base-catalyzed Knoevenagel condensation reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isophthalaldehyde is used in the synthesis of binuclear ruthenium complex. It participates in base-catalyzed Knoevenagel condensation reaction.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Store at room temperature. Keep away from oxidizing agents.
Isophthalaldehyde is used in the synthesis of binuclear ruthenium complex. It participates in base-catalyzed Knoevenagel condensation reaction.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Store at room temperature. Keep away from oxidizing agents.
Specifications
Specifications
| Melting Point | 85°C to 91°C |
| Boiling Point | 136°C (17 mmHg) |
| Quantity | 5 g |
| Beilstein | 1561038 |
| Sensitivity | Air Sensitive |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 134.13 |
| Percent Purity | 98% |
| Chemical Name or Material | Isophthalaldehyde |
Safety and Handling
| EINECSNumber | 210-935-8 |
| RTECSNumber | NT1981000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only