missing translation for 'onlineSavingsMsg'
Learn More

Isonitrosoacetophenone 98.0+%, TCI America™

Catalog No. P018110G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P018110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P018110G Supplier TCI America Supplier No. P018110G
Only null left

Chemical Identifiers

CAS 532-54-7
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.149
MDL Number MFCD00002122
InChI Key MLNKXLRYCLKJSS-RMKNXTFCSA-N
Synonym isonitrosoacetophenone, 2-isonitrosoacetophenone, benzoylformaldoxime, inaf, phenylglyoxal aldoxime, phenylglyoxal 2-oxime, 2-hydroxyiminoacetophenone, phenylglyoxal monoxime, acetophenone, 2-hydroxyimino, phenylglyoxaldoxime
PubChem CID 9566037
IUPAC Name (2E)-2-hydroxyimino-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)C=NO

Specifications

Melting Point 128°C
Quantity 10 g
Formula Weight 149.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isonitrosoacetophenone
RTECSNumber MD3325000
TSCA Yes