accessibility menu, dialog, popup

UserName

HONO(E)

CAS RN 532-54-7

IUPAC Name: (2E)-2-(N-hydroxyimino)-1-phenylethan-1-one
Synonyms: isonitrosoacetophenone
Molecular Weight (g/mol): 149.15
Molecular Formula: C8H7NO2
InChi Key: MLNKXLRYCLKJSS-RMKNXTFCSA-N
SMILES: O\N=C\C(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results