missing translation for 'onlineSavingsMsg'
Learn More

Isonicotinic acid hydrazide, 98+%

CAS: 54-85-3 | C6H7N3O | 137.142 g/mol

Supplier:  Thermo Scientific Chemicals A105830B

 View more versions of this product

Catalog No. AAA105830B

Only null left
Explore available promotions

Description

Description

Used for fluorescent HPLC detection of delta 4-3-ketosteroids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Isonicotinic acid hydrazide
169°C to 174°C
MFCD00006426
119374
14,5186
QRXWMOHMRWLFEY-UHFFFAOYSA-N
pyridine-4-carbohydrazide
3767
137.14
54-85-3
C6H7N3O
1000 g
Air Sensitive
isoniazid, isonicotinic acid hydrazide, isonicotinohydrazide, rimifon, isoniazide, nydrazid, isonicotinic hydrazide, laniazid, isonicotinylhydrazine, andrazide
C1=CN=CC=C1C(=O)NN
137.142
CHEBI:6030
≥98%

Chemical Identifiers

54-85-3
137.142
QRXWMOHMRWLFEY-UHFFFAOYSA-N
3767
pyridine-4-carbohydrazide
C6H7N3O
MFCD00006426
isoniazid, isonicotinic acid hydrazide, isonicotinohydrazide, rimifon, isoniazide, nydrazid, isonicotinic hydrazide, laniazid, isonicotinylhydrazine, andrazide
CHEBI:6030
C1=CN=CC=C1C(=O)NN
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 200-214-6

RTECSNumber : NS1751850

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only