missing translation for 'onlineSavingsMsg'
Learn More

Isonicotinic acid hydrazide, 98+%

Catalog No. p-7080859
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
1000 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1058318 50 g
AAA1058322 100 g
AAA1058336 500 g
AAA105830B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1058318 Supplier Thermo Scientific Chemicals Supplier No. A1058318
Only null left

CAS: 54-85-3 | C6H7N3O | 137.142 g/mol

Used for fluorescent HPLC detection of delta 4-3-ketosteroids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54-85-3
Molecular Formula C6H7N3O
Molecular Weight (g/mol) 137.142
MDL Number MFCD00006426
InChI Key QRXWMOHMRWLFEY-UHFFFAOYSA-N
Synonym isoniazid, isonicotinic acid hydrazide, isonicotinohydrazide, rimifon, isoniazide, nydrazid, isonicotinic hydrazide, laniazid, isonicotinylhydrazine, andrazide
PubChem CID 3767
ChEBI CHEBI:6030
IUPAC Name pyridine-4-carbohydrazide
SMILES C1=CN=CC=C1C(=O)NN

Specifications

Melting Point 169°C to 174°C
Quantity 50 g
Beilstein 119374
Sensitivity Air Sensitive
Merck Index 14,5186
Formula Weight 137.14
Percent Purity ≥98%
Chemical Name or Material Isonicotinic acid hydrazide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 200-214-6
RTECSNumber NS1751850
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only