accessibility menu, dialog, popup

UserName

CAS RN 54-85-3

IUPAC Name: pyridine-4-carbohydrazide
Synonyms: isoniazid nicizina tubazid tebecid niadrin nydrazid zinadon zonazide mybasan ismazide
Molecular Weight (g/mol): 137.14
Molecular Formula: C6H7N3O
InChi Key: QRXWMOHMRWLFEY-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=NC=C1
Molecular Weight (g/mol) 
  • (3)
  • (7)
Percent Purity 
  • (4)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (3)
  • (7)
  • (3)
  • (7)
  • (4)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)