missing translation for 'onlineSavingsMsg'
Learn More

Isoeugenol (cis- and trans- mixture) 97.0+%, TCI America™

Catalog No. p-7131225
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I013225G 25 g
I0132500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I013225G Supplier TCI America Supplier No. I013225G
Only null left

Chemical Identifiers

CAS 97-54-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00009285
InChI Key BJIOGJUNALELMI-ONEGZZNKSA-N
Synonym isoeugenol, e-isoeugenol, trans-isoeugenol, 2-methoxy-4-propenylphenol, 4-propenylguaiacol, 4-hydroxy-3-methoxy-1-propenylbenzene, 2-methoxy-4-prop-1-en-1-yl phenol, trans-p-propenylquaiacol, isoeugenol trans-form, 4-hydroxy-3-methoxypropenylbenzene
PubChem CID 853433
ChEBI CHEBI:50545
IUPAC Name 2-methoxy-4-[(1E)-prop-1-en-1-yl]phenol
SMILES COC1=CC(\C=C\C)=CC=C1O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 164.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoeugenol (cis- and trans- mixture)
EINECSNumber (3)-0637
RTECSNumber SL7875000
TSCA Yes