accessibility menu, dialog, popup

UserName

H3COCH3HO(E)

CAS RN 97-54-1

IUPAC Name: 2-methoxy-4-[(1E)-prop-1-en-1-yl]phenol
Synonyms: isoeugenol iso-eugenol 2 isoeugenol (I) trans-p-propenylquaiacol isoeugenol trans-form isoeugenol Z
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: BJIOGJUNALELMI-ONEGZZNKSA-N
SMILES: COC1=CC(\C=C\C)=CC=C1O
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (3)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)