missing translation for 'onlineSavingsMsg'
Learn More

Isoeugenol, cis + trans, 98%

Catalog No. AAB2454136
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2454136 500 g
AAB2454114 25 g
AAB2454122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2454136 Supplier Thermo Scientific Chemicals Supplier No. B2454136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 97-54-1 | C10H12O2 | 164.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97-54-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00009285
InChI Key BJIOGJUNALELMI-ONEGZZNKSA-N
Synonym isoeugenol, e-isoeugenol, trans-isoeugenol, 2-methoxy-4-propenylphenol, 4-propenylguaiacol, 4-hydroxy-3-methoxy-1-propenylbenzene, 2-methoxy-4-prop-1-en-1-yl phenol, trans-p-propenylquaiacol, isoeugenol trans-form, 4-hydroxy-3-methoxypropenylbenzene
PubChem CID 853433
ChEBI CHEBI:50545
IUPAC Name 2-methoxy-4-[(E)-prop-1-enyl]phenol
SMILES COC1=CC(\C=C\C)=CC=C1O

Specifications

Melting Point -10°C
Density 1.08
Boiling Point 266°C to 268°C
Flash Point 134°C (273°F)
Refractive Index 1.577
Quantity 500 g
Beilstein 1909602
Merck Index 14,5171
Formula Weight 164.21
Percent Purity ≥98%
Chemical Name or Material Isoeugenol, cis + trans
Show More Show Less
Hazard Category H302-H315-H317-H319
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P362-P501c
EINECSNumber 202-590-7
RTECSNumber SL7875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only