missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoeugenol, 98+%, mixture of cis/trans isomers
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 19.0°C to 23.0°C |
| Color | Yellow |
| Density | 1.0900g/mL |
| Boiling Point | 266.0°C to 268.0°C |
| Flash Point | 150°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | CH3OC6H3(CH=CHCH3)OH |
| Refractive Index | 1.5760 to 1.5780 |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | GHS Signal Word: Warning |
| missing translation for 'healthHazard2' | GHS H Statement Causes skin irritation. Harmful if swallowed. May cause an allergic skin reaction. Causes serious eye irritation. |
| missing translation for 'healthHazard3' | GHS P Statement IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse |
| EINECSNumber | 202-590-7 |
| RTECSNumber | SL7875000 |
| TSCA | TSCA |
RUO – Research Use Only