missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Isocyanoimino)triphenylphosphorane 95.0+%, TCI America™
Supplier: TCI America I064325G
Specifications
| (Isocyanoimino)triphenylphosphorane | |
| 152°C | |
| MFCD09038528 | |
| isocyanoimino triphenylphosphorane, isocyanoiminotriphenylphosphorane, isocyano triphenyl-??-phosphanylidene amine, cnnpph3;, ncnpph3;, ph3pnnc;, acmc-209os9, cn-n=pph3;, n-isocyaniminotriphenylphosphorane, n-isocyanoiminotriphenylphosphorane | |
| [C-]#[N+]N=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 302.32 | |
| 302.32 | |
| Crystalline Powder |
| 73789-56-7 | |
| C19H15N2P | |
| 25 g | |
| NIDTXBFHPXMXTR-UHFFFAOYSA-N | |
| isocyano(triphenyl-λâµ-phosphanylidene)amine | |
| 11266621 | |
| ≥95.0% (HPLC) |
Chemical Identifiers
| 73789-56-7 | |
| 302.32 | |
| NIDTXBFHPXMXTR-UHFFFAOYSA-N | |
| 11266621 | |
| [C-]#[N+]N=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C19H15N2P | |
| MFCD09038528 | |
| isocyanoimino triphenylphosphorane, isocyanoiminotriphenylphosphorane, isocyano triphenyl-??-phosphanylidene amine, cnnpph3;, ncnpph3;, ph3pnnc;, acmc-209os9, cn-n=pph3;, n-isocyaniminotriphenylphosphorane, n-isocyanoiminotriphenylphosphorane | |
| isocyano(triphenyl-λâµ-phosphanylidene)amine |
Safety and Handling
TSCA : No