accessibility menu, dialog, popup

UserName

CAS RN 73789-56-7

IUPAC Name: isocyano(triphenyl-λ⁵-phosphanylidene)amine
Synonyms: isocyano(triphenyl-λ⁵-phosphanylidene)amine
Molecular Weight (g/mol): 302.32
Molecular Formula: C19H15N2P
InChi Key: NIDTXBFHPXMXTR-UHFFFAOYSA-N
SMILES: [C-]#[N+]N=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)