missing translation for 'onlineSavingsMsg'
Learn More

(Isocyanoimino)triphenylphosphorane, 90%

Catalog No. AC452920010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC452920010 1 g
AC452920050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC452920010 Supplier Thermo Scientific Chemicals Supplier No. 452920010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 73789-56-7 | C19H15N2P | 302.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 73789-56-7
Molecular Formula C19H15N2P
Molecular Weight (g/mol) 302.32
MDL Number MFCD09038528
InChI Key NIDTXBFHPXMXTR-UHFFFAOYSA-N
Synonym isocyanoimino triphenylphosphorane, isocyanoiminotriphenylphosphorane, isocyano triphenyl-??-phosphanylidene amine, cnnpph3;, ncnpph3;, ph3pnnc;, acmc-209os9, cn-n=pph3;, n-isocyaniminotriphenylphosphorane, n-isocyanoiminotriphenylphosphorane
PubChem CID 11266621
IUPAC Name isocyanoimino(triphenyl)-$l^{5}-phosphane
SMILES [C-]#[N+]N=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 160°C to 161°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (HPLC)
Quantity 1 g
Formula Weight 302.31
Percent Purity 90%
Chemical Name or Material (Isocyanoimino)triphenylphosphorane
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only