Learn More
Isobutyronitrile, 99%
CAS: 78-82-0 | C4H7N | 69.107 g/mol
$155.18 - $155.18
Chemical Identifiers
| CAS | 78-82-0 |
|---|---|
| Molecular Formula | C4H7N |
| Molecular Weight (g/mol) | 69.107 |
| MDL Number | MFCD00001873 |
| InChI Key | LRDFRRGEGBBSRN-UHFFFAOYSA-N |
| Synonym | isobutyronitrile, isopropyl cyanide, propanenitrile, 2-methyl, 2-methylpropionitrile, isopropylnitrile, 2-cyanopropane, dimethylacetonitrile, isopropyl nitrile, 1-cyano-1-methylethane, isopropylkyanid |
| PubChem CID | 6559 |
| ChEBI | CHEBI:28638 |
| IUPAC Name | 2-methylpropanenitrile |
| SMILES | CC(C)C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL14470AU
|
Thermo Scientific Chemicals
L14470AU |
1000 mL |
Each for $155.18
|
|
|||||
Description
Used in the synthesis of gasoline additives, as a catalyst in the production of polyethylene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed in the synthesis of gasoline additives, as a catalyst in the production of polyethylene.
Solubility
Soluble in water 35g/L (20°C).
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
| 78-82-0 | |
| 69.107 | |
| LRDFRRGEGBBSRN-UHFFFAOYSA-N | |
| 6559 | |
| 2-methylpropanenitrile |
| C4H7N | |
| MFCD00001873 | |
| isobutyronitrile, isopropyl cyanide, propanenitrile, 2-methyl, 2-methylpropionitrile, isopropylnitrile, 2-cyanopropane, dimethylacetonitrile, isopropyl nitrile, 1-cyano-1-methylethane, isopropylkyanid | |
| CHEBI:28638 | |
| CC(C)C#N |
Specifications
| 78-82-0 | |
| 0.77 | |
| 8°C (46°F) | |
| 1.374 | |
| MFCD00001873 | |
| UN2284 | |
| 14,5156 | |
| Soluble in water 35g/L (20°C). | |
| CC(C)C#N | |
| 69.107 | |
| CHEBI:28638 | |
| 99% |
| -72°C | |
| 103°C to 104°C | |
| C4H7N | |
| (CH3)2CHCN | |
| 1000 mL | |
| 1340512 | |
| isobutyronitrile, isopropyl cyanide, propanenitrile, 2-methyl, 2-methylpropionitrile, isopropylnitrile, 2-cyanopropane, dimethylacetonitrile, isopropyl nitrile, 1-cyano-1-methylethane, isopropylkyanid | |
| LRDFRRGEGBBSRN-UHFFFAOYSA-N | |
| 2-methylpropanenitrile | |
| 6559 | |
| 69.11 | |
| Isobutyronitrile |
Safety and Handling
GHS H Statement
H225-H301-H311-H330
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P309+P311-P310-P312-P330-P363-P370+P378q-P501c
H225-H300+H330-H311-H371
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOBUTYRONITRILE
EINECSNumber : 201-147-5
RTECSNumber : TZ4900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only