missing translation for 'onlineSavingsMsg'
Learn More

Isobutyronitrile, 99%

CAS: 78-82-0 | C4H7N | 69.107 g/mol

Supplier:  Thermo Scientific Chemicals L14470AU

 View more versions of this product

Catalog No. AAL14470AU


Only null left
Explore available promotions

Description

Description

Used in the synthesis of gasoline additives, as a catalyst in the production of polyethylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of gasoline additives, as a catalyst in the production of polyethylene.

Solubility
Soluble in water 35g/L (20°C).

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

Isobutyronitrile
-72°C
103°C to 104°C
C4H7N
(CH3)2CHCN
1000 mL
1340512
isobutyronitrile, isopropyl cyanide, propanenitrile, 2-methyl, 2-methylpropionitrile, isopropylnitrile, 2-cyanopropane, dimethylacetonitrile, isopropyl nitrile, 1-cyano-1-methylethane, isopropylkyanid
LRDFRRGEGBBSRN-UHFFFAOYSA-N
2-methylpropanenitrile
6559
69.11
78-82-0
0.77
8°C (46°F)
1.374
MFCD00001873
UN2284
14,5156
Soluble in water 35g/L (20°C).
CC(C)C#N
69.107
CHEBI:28638
99%

Chemical Identifiers

78-82-0
69.107
LRDFRRGEGBBSRN-UHFFFAOYSA-N
6559
2-methylpropanenitrile
C4H7N
MFCD00001873
isobutyronitrile, isopropyl cyanide, propanenitrile, 2-methyl, 2-methylpropionitrile, isopropylnitrile, 2-cyanopropane, dimethylacetonitrile, isopropyl nitrile, 1-cyano-1-methylethane, isopropylkyanid
CHEBI:28638
CC(C)C#N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H301-H311-H330
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P309+P311-P310-P312-P330-P363-P370+P378q-P501c

H225-H300+H330-H311-H371

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOBUTYRONITRILE

EINECSNumber : 201-147-5

RTECSNumber : TZ4900000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only