missing translation for 'onlineSavingsMsg'
Learn More

Isobutyric anhydride, 97%

CAS: 97-72-3 | C8H14O3 | 158.197 g/mol

Supplier:  Thermo Scientific Chemicals L132400F

 View more versions of this product

Catalog No. AAL132400F


Only null left
Explore available promotions

Description

Description

Isobutyric anhydride is used in the preparation of 4-O-isobutyryl derivative by reacting with octyl beta-D-glucopyranoside using C2-symmetric chiral 4-pyrrolidinopyridine as a catalyst. It acts as a starting material for esters like sucrose acetate isobutyrate, dyes and agrochemicals. It finds application as fragrance intermediate and flavorings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobutyric anhydride is used in the preparation of 4-O-isobutyryl derivative by reacting with octyl beta-D-glucopyranoside using C2-symmetric chiral 4-pyrrolidinopyridine as a catalyst. It acts as a starting material for esters like sucrose acetate isobutyrate, dyes and agrochemicals. It finds application as fragrance intermediate and flavorings.

Solubility
Miscible with water and ether.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

Isobutyric anhydride
-56°C
182°C
Pungent
1.406
2500 mL
386267
isobutyric anhydride, isobutyric acid anhydride, propanoic acid, 2-methyl-, anhydride, 2-methylpropanoic anhydride, isobutyricanhydride, isobutyryl anhydride, 2-methylpropionic anhydride, unii-n85a80fjdt, n85a80fjdt, 2-methylpropanoic acid anhydride
LSACYLWPPQLVSM-UHFFFAOYSA-N
2-methylpropanoyl 2-methylpropanoate
7346
158.2
97-72-3
0.954
67°C (152°F)
C8H14O3
MFCD00008913
UN2922
Moisture sensitive
Miscible with water and ether.
CC(C)C(=O)OC(=O)C(C)C
158.197
CHEBI:84261
97%

Chemical Identifiers

97-72-3
158.197
LSACYLWPPQLVSM-UHFFFAOYSA-N
7346
2-methylpropanoyl 2-methylpropanoate
C8H14O3
MFCD00008913
isobutyric anhydride, isobutyric acid anhydride, propanoic acid, 2-methyl-, anhydride, 2-methylpropanoic anhydride, isobutyricanhydride, isobutyryl anhydride, 2-methylpropionic anhydride, unii-n85a80fjdt, n85a80fjdt, 2-methylpropanoic acid anhydride
CHEBI:84261
CC(C)C(=O)OC(=O)C(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.

P210-P233-P240-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P363-P374-P501c

H227-H311+H331-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

EINECSNumber : 202-603-6

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only