accessibility menu, dialog, popup

UserName

CAS RN 97-72-3

IUPAC Name: 2-methylpropanoyl 2-methylpropanoate
Synonyms: 2-methylpropanoyl 2-methylpropanoate
Molecular Weight (g/mol): 158.2
Molecular Formula: C8H14O3
InChi Key: LSACYLWPPQLVSM-UHFFFAOYSA-N
SMILES: CC(C)C(=O)OC(=O)C(C)C
Boiling Point 
  • (5)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)