missing translation for 'onlineSavingsMsg'
Learn More

Isobutyric anhydride, 97%

Catalog No. p-7046130
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13240AE 100 mL
AAL13240AP 500 mL
AAL132400F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL13240AE Supplier Thermo Scientific Chemicals Supplier No. L13240AE
Only null left

CAS: 97-72-3 | C8H14O3 | 158.197 g/mol

Isobutyric anhydride is used in the preparation of 4-O-isobutyryl derivative by reacting with octyl beta-D-glucopyranoside using C2-symmetric chiral 4-pyrrolidinopyridine as a catalyst. It acts as a starting material for esters like sucrose acetate isobutyrate, dyes and agrochemicals. It finds application as fragrance intermediate and flavorings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobutyric anhydride is used in the preparation of 4-O-isobutyryl derivative by reacting with octyl beta-D-glucopyranoside using C2-symmetric chiral 4-pyrrolidinopyridine as a catalyst. It acts as a starting material for esters like sucrose acetate isobutyrate, dyes and agrochemicals. It finds application as fragrance intermediate and flavorings.

Solubility
Miscible with water and ether.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 97-72-3
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD00008913
InChI Key LSACYLWPPQLVSM-UHFFFAOYSA-N
Synonym isobutyric anhydride, isobutyric acid anhydride, propanoic acid, 2-methyl-, anhydride, 2-methylpropanoic anhydride, isobutyricanhydride, isobutyryl anhydride, 2-methylpropionic anhydride, unii-n85a80fjdt, n85a80fjdt, 2-methylpropanoic acid anhydride
PubChem CID 7346
ChEBI CHEBI:84261
IUPAC Name 2-methylpropanoyl 2-methylpropanoate
SMILES CC(C)C(=O)OC(=O)C(C)C

Specifications

Melting Point -56°C
Density 0.954
Boiling Point 182°C
Flash Point 67°C (152°F)
Odor Pungent
Refractive Index 1.406
Quantity 100 mL
UN Number UN2922
Beilstein 386267
Sensitivity Moisture sensitive
Solubility Information Miscible with water and ether.
Formula Weight 158.2
Percent Purity 97%
Chemical Name or Material Isobutyric anhydride
Show More Show Less
Hazard Category H227-H311+H331-H314
Hazard Statement GHS H Statement
H314-H318-H302-H312-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
Precautionary Statement P210-P233-P240-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P363-P374-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 202-603-6
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only